CAS RN 125068-54-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSP-805, MedChemExpress
MedChemExpress NSP-805 is a potent and selective inhibitor of guinea pig cardiac phosphodiesterase 3 (PDE3), and a cardiotonic agent with vasodilator properties.