CAS RN 124555-18-6
IUPAC Nombre:
(2S,3S,4S,5R)-2-{6-[(2-hydroxycyclopentyl)amino]-9H-purin-9-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Sinónimos:
(2S,3S,4S,5R)-2-{6-[(2-hydroxycyclopentyl)amino]purin-9-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Peso molecular (g/mol):
351.36
Fórmula molecular:
C15H21N5O5
InChi Key:
GYWXTRVEUURNEW-FPHQEEKISA-N
SMILES:
OC[C@H]1O[C@@H]([C@@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCC3O)N=CN=C12
1
–
1
de
1
resultados
1
GR79236, MedChemExpress
MedChemExpress GR79236 is a highly potent, selective and orally active adenosine A1 receptor agonist with a Kis of 3.1 nM and 1300 nM for A1 and A2 receptors, respectively. GR79236 has anti-nociceptive and anti-inflammatory actions.