CAS RN 1245319-54-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PI4KIII beta inhibitor 3, MedChemExpress
MedChemExpress PI4KIII beta inhibitor 3 is a novel and high effective PI4KIIIβ inhibitor with IC50 of 5.7 nM.