CAS RN 1243245-18-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GNF-6231, MedChemExpress
MedChemExpress GNF-6231 is a potent, selective, and orally bioavailable Porcupine inhibitor that blocks Wnt signaling.