CAS RN 1235449-52-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BEBT-908, MedChemExpress
MedChemExpress BEBT-908 (PI3Kα inhibitor 1) is a selective PI3Kα inhibitor extracted from patent US/20120088764A1, Compound 243, has an IC5050≤1 μM) .