CAS RN 1227633-49-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ StemRegenin 1
Aryl hydrocarbon receptor (AhR) antagonist; promotes HSC expansion and engraftment
2
StemRegenin 1, MedChemExpress
MedChemExpress StemRegenin 1 is a potent aryl hydrocarbon receptor (AhR) antagonist with IC50 of 127 nM.
3