CAS RN 122341-56-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress AMPPD
MedChemExpress AMPPD, a 1,2-dioxo-cyclohexane derivative, is a biochemistry ultrasensitive alkaline phosphatase substrate.
2
MedChemExpress AMPPD
MedChemExpress AMPPD, a 1,2-dioxo-cyclohexane derivative, is a biochemistry ultrasensitive alkaline phosphatase substrate.
3
MedChemExpress AMPPD
MedChemExpress AMPPD, a 1,2-dioxo-cyclohexane derivative, is a biochemistry ultrasensitive alkaline phosphatase substrate.
4
MedChemExpress AMPPD
MedChemExpress AMPPD, a 1,2-dioxo-cyclohexane derivative, is a biochemistry ultrasensitive alkaline phosphatase substrate.