CAS RN 122341-40-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Neuropeptide Y (13-36), amide, human
MedChemExpress Neuropeptide Y (13-36), amide, human is a selective neuropeptide Y2 receptor agonist.
2
MedChemExpress Neuropeptide Y (13-36), amide, human
MedChemExpress Neuropeptide Y (13-36), amide, human is a selective neuropeptide Y2 receptor agonist.