CAS RN 1221443-94-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
A-1165442, MedChemExpress
MedChemExpress A-1165442 is a potent, competitive and orally available TRPV1 antagonist with an IC50 of 9 nM for human TRPV1.