CAS RN 122063-01-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress [bAla8]-Neurokinin A(4-10)
MedChemExpress [bAla8]-Neurokinin A(4-10) is a neurokinin 2 (NK2) receptor agonist.
2
MedChemExpress [bAla8]-Neurokinin A(4-10)
MedChemExpress [bAla8]-Neurokinin A(4-10) is a neurokinin 2 (NK2) receptor agonist.
3
MedChemExpress [bAla8]-Neurokinin A(4-10)
MedChemExpress [bAla8]-Neurokinin A(4-10) is a neurokinin 2 (NK2) receptor agonist.