CAS RN 1217471-51-6
IUPAC Nombre:
2-({5-[(2-amino-3-sulfanylpropyl)amino]-[1,1'-biphenyl]-2-yl}formamido)-4-(methylsulfanyl)butanoic acid; trifluoroacetic acid
Sinónimos:
2-({5-[(2-amino-3-sulfanylpropyl)amino]-[1,1'-biphenyl]-2-yl}formamido)-4-(methylsulfanyl)butanoic acid; trifluoroacetic acid
Peso molecular (g/mol):
547.61
Fórmula molecular:
C23H28F3N3O5S2
InChi Key:
IAVHISGQCODLHL-UHFFFAOYNA-N
SMILES:
OC(=O)C(F)(F)F.CSCCC(NC(=O)C1=C(C=C(NCC(N)CS)C=C1)C1=CC=CC=C1)C(O)=O
1
–
1
de
1
resultados
1
FTI 276 TFA, MedChemExpress
MedChemExpress FTI-276 is a protein farnesyl transferase (PFT) inhibitor with IC50s of 0.9 nM and 0.5 nM for Plasmodium falciparum and human, respectively.