CAS RN 1215833-62-7
IUPAC Nombre:
methyl 4-ethyl-6,7-dimethoxy-9H-pyrido[3,4-b]indole-3-carboxylate hydrochloride
Sinónimos:
DMCM hydrochloride
Peso molecular (g/mol):
350.8
Fórmula molecular:
C17H19ClN2O4
InChi Key:
FHCRBVQGMZUJKY-UHFFFAOYSA-N
SMILES:
Cl.CCC1=C(N=CC2=C1C1=C(N2)C=C(OC)C(OC)=C1)C(=O)OC
1
–
2
de
2
resultados
1
Tocris Bioscience™ DMCM hydrochloride
CAS: 1215833-62-7 Fórmula molecular: C17H19ClN2O4
2
DMCM hydrochloride, MedChemExpress
MedChemExpress DMCM hydrochloride is a nonselective full inverse agonist of benzodiazepine. DMCM shows bnding afinity at human recombinant GABAA αxβ3γ2 receptor subtypes with Kis of 10 nM, 13 nM, 7.5 nM, 2.2 nM for α1, α2, α3, and α5 receptors, respectively.