CAS RN 1208243-50-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Cibinetide
MedChemExpress Cibinetide (ARA290) is an EPO-derivative, acting as a specific agonist of erythropoietin/CD131 heteroreceptor, and used for neurological disease treatment.
2
MedChemExpress Cibinetide
MedChemExpress Cibinetide (ARA290) is an EPO-derivative, acting as a specific agonist of erythropoietin/CD131 heteroreceptor, and used for neurological disease treatment.
3
MedChemExpress Cibinetide
MedChemExpress Cibinetide (ARA290) is an EPO-derivative, acting as a specific agonist of erythropoietin/CD131 heteroreceptor, and used for neurological disease treatment.