CAS RN 1208123-85-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CaMKII-IN-1, MedChemExpress
MedChemExpress CaMKII-IN-1 is a potent and highly selective CaMKII inhibitor with IC50 of 63 nM; significantly high selectivity against CaMKIV, MLCK, p38a, Akt1, and PKC.