CAS RN 1206711-16-1
IUPAC Nombre:
4-{6-[4-(propan-2-yloxy)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl}quinoline
Sinónimos:
4-[6-(4-isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline
Peso molecular (g/mol):
380.45
Fórmula molecular:
C24H20N4O
InChi Key:
JMIFGARJSWXZSH-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=C2C=CC=CC2=NC=C1
1
–
4
de
4
resultados
1
Tocris Biosciences™ DMH-1
Selective ALK2 inhibitor
2
Tocris Bioscience™ DMH-1
CAS: 1206711-16-1 Fórmula molecular: C24H20N4O
3
DMH-1, MedChemExpress
MedChemExpress DMH-1 is a potent and selective BMP inhibitor with IC50s of 27/107.9/<5/47.6 nM for ALK1/ALK2/ALK3/ALK6, respectively.
4