CAS RN 1206101-20-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GLPG0634 analog, MedChemExpress
MedChemExpress GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.