CAS RN 1202710-57-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSTO-IN-2, MedChemExpress
MedChemExpress GSTO-IN-2 is a glutathione S-transferase inhibitor with IC50s of 3.6, 16.3, and 1.4 μM for GSTA2, GSTM1, and GSTP1-1.