CAS RN 1198786-98-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
INT-777 R-enantiomer, MedChemExpress
MedChemExpress INT-777 (R-enantiomer) is the R-enantiomer of INT-777, with EC50 of 4.79 μM for TGR5, and less potent than INT-777.