CAS RN 1196541-47-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GDC-0575, MedChemExpress
MedChemExpress GDC-0575 (ARRY-575, RG7741) is a highly-selective oral small-molecule Chk1 inhibitor with an IC50 of 1.2 nM.