CAS RN 1185242-90-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Ácido micofenólico-d3, TRC
CAS: 1185242-90-3 Fórmula molecular: C17 2H3 H17 O6 Peso molecular (g/mol): 323.36 Sinónimo: Mycophenolic Acid-(methoxy-D3) Nombre IUPAC: (E)-6-[4-hidroxi-7-metil-3-oxo-6-(trideuteriometoxi)-1H-2-benzofurano-5-il]-4-metilhex-4-ácido enoico SMILES: [2H]C([2H])([2H])Oc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)O
2
Mycophenolic acid-d3, MedChemExpress
MedChemExpress Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity.