CAS RN 1184136-10-4
IUPAC Nombre:
1-(3-bromobenzoyl)-4-(pyrrolidin-1-yl)piperidine hydrochloride
Sinónimos:
1-(3-bromobenzoyl)-4-(pyrrolidin-1-yl)piperidine hydrochloride
Peso molecular (g/mol):
373.72
Fórmula molecular:
C16H22BrClN2O
InChi Key:
DPPMBBHOXQOMJI-UHFFFAOYSA-N
SMILES:
Cl.BrC1=CC=CC(=C1)C(=O)N1CCC(CC1)N1CCCC1
1
–
1
de
1
resultados
1
UNC926, MedChemExpress
MedChemExpress UNC926 is a methyl-lysine (Kme) reader domain inhibitor that inhibits L3MBTL1 with an IC50 of 3.9 μM.