CAS RN 1182367-47-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
RAD 140, TRC
CAS: 1182367-47-0 Fórmula molecular: C20H16ClN5O2 Peso molecular (g/mol): 393.8 Sinónimo: Testolone Nombre IUPAC: 2-cloro-4-[[(1R,2S)-1-[5-(4-cianofenil)-1,3,4-oxadiazol-2-il]-2-hidroxipropil]amino-3-metil-benzonitrilo SMILES: C[C@H](O)[C@](NC1=C(C)C(Cl)=C(C#N)C=C1)([H])C2=NN=C(C3=CC=C(C#N)C=C3)O2
2
Vosilasarm, MedChemExpress
MedChemExpress Vosilasarm (RAD140) is a potent, orally active, nonsteroidal selective androgen receptor modulator (SARM) with a Ki of 7 nM. Vosilasarm shows good selectivity over other steroid hormone nuclear receptors.