CAS RN 1172637-87-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
gamma-secretase modulator 1, MedChemExpress
MedChemExpress γ-secretase inhibitior-1 is a gamma-secretase modulator, γ-secretase inhibitior-1 is useful for Alzheimer's disease.