CAS RN 116209-55-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Levobetaxolol hydrochloride, MedChemExpress
MedChemExpress Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor (beta blocker) that can lower the pressure in the eye.