CAS RN 116-38-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Edrophonium chloride, MedChemExpress
MedChemExpress Edrophonium chloride is a readily reversible acetylcholinesterase inhibitor; prevents breakdown of the neurotransmitter acetylcholine and acts by competitively inhibiting the enzyme acetylcholinesterase, mainly at the neuromuscular junction.