CAS RN 1159840-61-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Gardiquimod diTFA, MedChemExpress
MedChemExpress Gardiquimod diTFA, an imidazoquinoline analog, is a TLR7/8 agonist. Gardiquimod diTFA could inhibit HIV-1 infection of macrophages and activated peripheral blood mononuclear cells (PBMCs). Gardiquimod diTFA specifically activates TLR7 when used at concentrations below 10 μM.