CAS RN 1158279-20-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HLCL-61 hydrochloride, MedChemExpress
MedChemExpress HLCL-61 hydrochloride is a first-in-class inhibitor of protein arginine methyltransferase 5 (PRMT5).