CAS RN 115810-12-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tubeimoside II, MedChemExpress
MedChemExpress Tubeimoside II(Tubeimoside-B) is a natural analogue of oleanane type of triterpenoid saponin; show anti-inflammatory, antitumor, and antitumor-promoting effects.