CAS RN 1146963-51-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
J-147, MedChemExpress
MedChemExpress J-147 is an exceptionally potent, orally active, neuroprotective agent for cognitive enhancement. J-147 can readily pass the blood brain barrier (BBB). J-147 can inhibit monoamine oxidase B (MAO B) and the dopamine transporter with EC50 values of 1.88 μM and 0.649 μM, respectively. J-147 has potential for the treatment of Alzheimer’s disease (AD).