CAS RN 1137608-69-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Telotristat etiprate, MedChemExpress
MedChemExpress Telotristat etiprate (LX1606 Hippurate) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.