CAS RN 1121-30-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pyrithione, MedChemExpress
MedChemExpress Pyrithione, a Transition metal complexe, is a zinc ionophore that causes increased zinc levels within mammalian cells. Pyrithione has potent bactericidal and anti-fungal activity.