CAS RN 110880-55-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress [Sar9,Met(O2)11]-Substance P
MedChemExpress [Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.
2
MedChemExpress [Sar9,Met(O2)11]-Substance P
MedChemExpress [Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.
3
MedChemExpress [Sar9,Met(O2)11]-Substance P
MedChemExpress [Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.
4
MedChemExpress [Sar9,Met(O2)11]-Substance P
MedChemExpress [Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.