CAS RN 110-49-6
IUPAC Nombre:
2-methoxyethyl acetate
Sinónimos:
2-methoxyethyl acetate
Peso molecular (g/mol):
118.13
Fórmula molecular:
C5H10O3
InChi Key:
XLLIQLLCWZCATF-UHFFFAOYSA-N
SMILES:
COCCOC(C)=O
1
–
2
de
2
resultados
1
2-Acetato de Metoxietilo, TRC
CAS: 110-49-6 Fórmula molecular: C5 H10 O3 Peso molecular (g/mol): 118.13 Sinónimo: Ethanol, 2-methoxy-, acetate (6CI,7CI,8CI,9CI),1-Acetoxy-2-methoxyethane,2-Methoxyethanol acetate,2-Methoxyethyl acetate,Acetyl methyl cellosolve,Ethylene glycol acetate monomethyl ether,Ethylene glycol methyl ether acetate,Ethylene glycol monomethyl ether acetate,Glycol monomethyl ether acetate,MC acetate,Methyl Cellosolve acetate,Methyl glycol acetate,β-Methoxyethyl acetate Nombre IUPAC: acetato de 2-metoxietilo SMILES: COCCOC(=O)C