CAS RN 109292-46-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Arg-Gly-Asp-Cys
MedChemExpress Arg-Gly-Asp-Cys is the binding motif of fibronectin to cell adhesion molecules, and can inhibit platelet aggregation and fibrinogen binding.
2
MedChemExpress Arg-Gly-Asp-Cys
MedChemExpress Arg-Gly-Asp-Cys is the binding motif of fibronectin to cell adhesion molecules, and can inhibit platelet aggregation and fibrinogen binding.
3
MedChemExpress Arg-Gly-Asp-Cys
MedChemExpress Arg-Gly-Asp-Cys is the binding motif of fibronectin to cell adhesion molecules, and can inhibit platelet aggregation and fibrinogen binding.