CAS RN 1092788-83-4
IUPAC Nombre:
1-cyclopentyl-3-{1H-pyrrolo[2,3-b]pyridin-5-yl}-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Sinónimos:
1-cyclopentyl-3-{1H-pyrrolo[2,3-b]pyridin-5-yl}pyrazolo[3,4-d]pyrimidin-4-amine
Peso molecular (g/mol):
319.37
Fórmula molecular:
C17H17N7
InChi Key:
NVRXTLZYXZNATH-UHFFFAOYSA-N
SMILES:
NC1=C2C(=NC=N1)N(N=C2C1=CN=C2NC=CC2=C1)C1CCCC1
1
–
2
de
2
resultados
1
PP121, MedChemExpress
MedChemExpress PP121 is a multi-targeted kinase inhibitor with IC50s of 10, 60, 12, 14, 2 nM for mTOR, DNK-PK, VEGFR2, Src, PDGFR, respectively.
2
PP 121
Dual kinase inhibitor; inhibits PI 3-K family kinases