CAS RN 108963-70-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate (Spectrozyme PCa) is a chromogenic substrate.
2
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate (Spectrozyme PCa) is a chromogenic substrate.
3
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate
MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate (Spectrozyme PCa) is a chromogenic substrate.