CAS RN 1088543-62-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress AZP-531
MedChemExpress AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight.
2
MedChemExpress AZP-531
MedChemExpress AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight.
3
MedChemExpress AZP-531
MedChemExpress AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight.