CAS RN 1087708-38-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(+)-S-N-Hidrocloruro de metilcoclaurina, TRC
CAS: 1087708-38-0 Fórmula molecular: C18H21NO3 . HCl Peso molecular (g/mol): 335.8 Sinónimo: 1,2,3,4-Tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-7-isoquinolinol Hydrochloride (1:1),(S)-2-Methylcoclaurine Hydrochloride,N-Methyl-coclaurine Hydrochloride,(+)-(S)-N-Methylcoclaurine Hydrochloride,(+)-N-Methylcoclaurine Hydrochloride,N-Methylcoclaurine Hydrochloride,S-N-Methylcoclaurine Hydrochloride Nombre IUPAC: (1S)-1-[(4-hidroxifenil)metil]-6-metaxi-2-metil-3,4-dihidro-1H-isoquinolina-7-ol; Cloruro SMILES: Cl.COc1cc2CCN(C)[C@@H](Cc3ccc(O)cc3)c2cc1O