Learn More
Phenomenex™ Zebron™ ZB-WAXplus, GC Capillary Column 30 m x 0.25 mm x 0.25 μm
Product Code. 31747640 Shop All Phenomenex Products
Change view
Click to view available options
Length (Metric):
30 m
Unit Size:
Each
Product Code. Length (Metric) Unit Size
31747640 30 m Each
1 options
This item is not returnable. View return policy
Product Code. 31747640 Supplier Phenomenex™ Supplier No. 7HGG01311
Save Up to 50% on Phenomenex Products
Use promo code 27955
View all promotions
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
GC columns that deliver low bleed, excellent retention, and long column life for precise, reproducible results.
  • Capillary Column
TRUSTED_SUSTAINABILITY
Specifications
Particle Size None
Film Thickness 0.25 μm
Length (Metric) 30 m
Packing Material Fused Silica
USP Type G14, G15, G16, G20, G39, G47
Stationary Phase Proprietary
Type GC Column
Diameter (Metric) Inner 0.25 mm