Learn More
DWK Life Sciences WHEATON™ Staining Jars, 8-10 Slide Unit
Product Code. 10284922 Shop All DWK Life Sciences Products
Change view
Click to view available options
Slide Count:
8 Individual or 16 Back to Back
Unit Size:
Pack of 6
Product Code. Slide Count Unit Size
10284922 8 Individual or 16 Back to Back Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 10284922 Supplier DWK Life Sciences Supplier No. 900620
  • ONLINE EXCLUSIVE PRICE
    288.00€
    254.88€ / paquete de 6
    Ahorro 33.12€
    11% Descuento
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Staining Jars, 8 to 10 Slide Unit feature a wide top designed for convenient slide transfer, making them especially suitable for staining slides inscribed on one end. Manufactured from soda-lime glass, each jar holds 8 single 3 x 1 in. slides vertically or 16 back-to-back.
Featuring a wide top designed for convenience in transferring slides. Wheaton™ Staining Jars, 8-10 Slide Unit are especially suitable for staining slides that are inscribed on one end.
  • Wide top designed for convenience in transferring slides
  • Especially suitable for staining slides that are inscribed on one end
  • Holds 8 single 3 x 1 in. (75 x 25 mm) slides vertically or 16 slides back-to-back
  • Manufactured from soda-lime glass
  • Approximate inside bottom dimensions: 47 L x 26 W x 85 D mm
  • Includes cover
Specifications
Closure Included Closure Included
Closure Material Glass
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Material Soda Lime Glass
Type Staining Dish
Slide Count 8 Individual or 16 Back to Back
Dimensions (L x W) 1.6 x 1.02 in. (4.2 x 2.6 cm)
Length (English) 1.6 in.
Length (Metric) 47 mm
Width (English) 1.02 in.
Width (Metric) 26 mm
Height (Metric) 79 mm
Depth (English) Inside 3.3 in.
Depth (Metric) 79 mm
Depth (Metric) Inside 8.5 cm