Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm LVI Amber Vial Convenience Pack with Screw Cap Top Finish and Marking Patch
Product Code. 12415021 Shop All Wheaton Science Products Products
Change view
Click to view available options
Closure Color:
Black
Blue
Green
Natural
Yellow
Septum:
Blue PTFE, Silicone
PTFE
PTFE, Silicone
PTFE, Silicone, PTFE
Red PTFE, Silicone
Pre-slit:
No
Yes
Unit Size:
Pack of 100
Product Code. Closure Color Septum Pre-slit unitSize
12415021 Black Red PTFE, Silicone No Pack of 100
12405021 Natural PTFE, Silicone No Pack of 100
12435021 Blue PTFE, Silicone No Pack of 100
12445021 Yellow PTFE, Silicone No Pack of 100
12455021 Green PTFE, Silicone No Pack of 100
12475021 Black PTFE, Silicone, PTFE No Pack of 100
12495021 Black Blue PTFE, Silicone No Pack of 100
12415031 Black PTFE No Pack of 100
10716554 Natural PTFE, Silicone Yes Pack of 100
12435031 Black PTFE, Silicone Yes Pack of 100
10 options
This item is not returnable. View return policy
Product Code. 12415021
Brand: DWK Life Sciences W2253280201
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
LVI™ Vial with a sealed-in 300 μL glass limited volume insert, fused to vial to prevent needle damage. Made of Type I borosilicate glass with white ceramic write-on patch.
  • Inserts, clear vials, and amber vials are converted from USP Type I borosilicate glass tubing
  • Insert is fused to vial to prevent needle damage
  • Borosilicate glass vial with 300 μL glass limited volume insert
  • Manufacturing process precisely controls the depth and position of the insert relative to the bottom of the vial. This allows more consistent sample recovery from vial to vial and reduces the possibility of autosampler needle damage.
  • Insert has a slight flare at the top, providing a larger needle target area, which prevents needle damage
  • Available in clear and amber
  • Purchase vials and caps separately or together in convenience packs
  • Packed in a case of 100
  • Writing patch on all vials
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Pre-slit No
Volume (Metric) 0.3 mL
Septum Red PTFE, Silicone
Thread Style Screw
Volume (English) 0.1 dr.
Closure Material Polypropylene
Closure Type Screw Cap
Marking Spot Yes
Height (Metric) 32 mm