Learn More
DWK Life Sciences WHEATON™ Glass Staining Dishes for 50 Slides
Product Code. 10162862 Shop All DWK Life Sciences Products
Change view
Click to view available options
Includes:
Cover
Dish
Dish, Cover
Dish, Cover, Rack
Rack
Unit Size:
Pack of 3
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10162862 Rack Pack of 3
10306472 Dish, Cover, Rack Pack of 6
10623703 Dish, Cover Pack of 3
10779474 Dish Pack of 3
10254732 Cover Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10162862 Supplier DWK Life Sciences Supplier No. 900404
  • ONLINE EXCLUSIVE PRICE
    246.00€
    232.99€ / paquete de 3
    Ahorro 13.01€
    5% Descuento
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Glass Staining Dishes for 50 Slides are made from soda-lime glass and hold up to 50 standard slides. The stainless steel rack is nontarnishable, resists staining solutions, and features a hinged handle for easy slide insertion and removal.
  • Dish and cover are made from soda-lime glass
  • Holds up to 50 standard slides ( 3 x 1, 3 x 1.5, 3 x 2 in.)
  • Stainless steel rack is nontarnishable and withstands most staining solutions
  • Hinged handle allows easy insertion and removal of slides
  • Open-bottom rack design facilitates rapid immersion and drainage, reducing carryover
  • Slides can be placed singly, alternately straight across and diagonally, or back-to-back
  • Dimensions: 185 L x 88 W x 78 mm D
  • Available as complete unit or as individual components (dish, cover, rack)
Specifications
Closure Included No
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Silver
Material Stainless Steel
Type Stainless Steel Rack
For Use With (Equipment) Glass staining dish for 50 slides
Includes Rack
Slide Count 50 slides horizontally