Learn More
DWK Life Sciences WHEATON™ Replacement Screw Cap for Coplin Staining Jar 900520/900570, 58-400 Finish
Product Code. 10182862 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
5 to 10 Slide Staining Dish
Unit Size:
Pack of 6
Product Code. For Use With (Equipment) Unit Size
10182862 5 to 10 Slide Staining Dish Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 10182862 Supplier DWK Life Sciences Supplier No. 900522
  • ONLINE EXCLUSIVE PRICE
    79.50€
    72.35€ / paquete de 6
    Ahorro 7.15€
    8% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Replacement Screw Cap for Coplin Staining Jar 900520/900570, 58-400 Finish is a linerless white polypropylene cap designed to reduce solvent evaporation and spills. Compatible with WHEATON™ Coplin staining jars. Cap is not intended for use with slides in jar.
  • Replacement screw cap (58-400 finish) for Wheaton Coplin Staining Jars 900520 and 900570
  • Made of linerless white polypropylene to reduce solvent evaporation and spills during storage
  • Cap is not intended for use with slides in jar
  • Sold as 6 per case
  • Supplied by DWK Life Sciences under the Wheaton brand
Specifications
Closure Type Screw
Autoclavable Not Autoclavable
Color White
Material Polypropylene
Type Screw Cap
For Use With (Equipment) 5 to 10 Slide Staining Dish
Height (Metric) 70 mm