Learn More
Phenomenex™ Verex™ Vial Inserts, 6 mm, Flat Bottom, Glass
Product Code. 31745821 Shop All Phenomenex Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 500
Product Code. Color Unit Size
31745821 Clear Pack of 500
1 options
This item is not returnable. View return policy
Product Code. 31745821 Supplier Phenomenex™ Supplier No. AR0461052
Fecha estimada de envoi: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Accessories for use with chromatography vials and in various chromatography applications.
  • Analysis: Cleanliness and performance tested as standard
  • Certification: 100% defect-free
  • Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 12 mm
Material Glass
Pre-slit No
Sterility Sterile
Closure Size 6 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Insert