missing translation for 'onlineSavingsMsg'
Learn More

TRIS, 0.5M buffer soln., pH 7.8, Thermo Scientific™

Product Code. 15484419
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Product Code. Quantity unitSize
15484419 500 mL 500mL
15494419 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 15484419

Brand: Thermo Scientific Alfa Aesar J61944.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 0.5 M
pH 7.8
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material TRIS buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.