missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99%, Thermo Scientific Chemicals

Product Code. 11489666
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
Product Code. Quantity unitSize
11489666 1000 mL 1000mL
11499666 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 11489666

Brand: Thermo Scientific Alfa Aesar B20215.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

Solubility
Miscible with tetrahydrofuran, ether, isoporpylamine, hexane and methanol.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It decomposes slowly in the presence of moisture and rapidly in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9BO3
Molecular Weight (g/mol) 103.912
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34°C
Density 0.915
Boiling Point 67°C to 69°C
Flash Point −8°C (17°F)
Odor Odorless
Linear Formula (CH3O)3B
Refractive Index 1.361
Quantity 1000 mL
UN Number UN2416
Beilstein 1697939
Sensitivity Moisture sensitive
Merck Index 14,9712
Solubility Information Miscible with tetrahydrofuran,ether,isoporpylamine,hexane and methanol.
Formula Weight 103.92
Percent Purity 99%
Chemical Name or Material Trimethyl borate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl borate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : methanol, Trimethylborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.