Learn More
Triethoxy(isobutyl)silane, 95%, BLD Pharm™
Product Code. 30802713
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30802713 1 kg 1kg
30801285 500 g 500g
30793039 25 g 25g
30801212 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30802713 Supplier BLD Pharm Supplier No. BD1350711000g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 17980-47-1
Molecular Formula C10H24O3Si
MDL Number MFCD00069084
Quantity 1 kg
SMILES CC(C)C[Si](OCC)(OCC)OCC
Molecular Weight (g/mol) 220.38
Percent Purity 95%
Chemical Name or Material Triethoxy(isobutyl)silane