Learn More
ZNF250 Antibody, PerCP, R&D Systems™
Product Code. 18621652 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
Product Code. Unit Size
18621652 0.10mL
1 options
This item is not returnable. View return policy
Product Code. 18621652 Supplier Novus Biologicals Supplier No. NBP297823PCP
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
ZNF250 Polyclonal specifically detects ZNF250 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Formulation PBS
Primary or Secondary Primary
Product Type Antibody
Product Line Novus