missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(1-ethoxyvinyl)tin, 95%

Product Code. 10338893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Código de producto. Quantity Packaging unitSize
10338893 25 g Glass bottle 25g
10413063 1 g Glass bottle 1g
10557513 5 g Glass bottle 5g
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10338893

Marca: Thermo Scientific Acros 377710250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 97674-02-7 | C16H34OSn | 361.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 97674-02-7
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00010240
InChI Key HGXJOXHYPGNVNK-UHFFFAOYSA-N
Synonym tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl
PubChem CID 619414
IUPAC Name tributyl(1-ethoxyethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)OCC

Especificaciones

Color Colorless
Density 1.0690g/mL
Boiling Point 85.0°C to 86.0°C (0.1 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]SnC(CH2)OCH2CH3
Refractive Index 1.4750 to 1.4770
Quantity 25 g
Specific Gravity 1.069
Solubility Information Solubility in water: insoluble
Formula Weight 361.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tributyl(1-ethoxyvinyl)tin
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Tributyl(1-ethoxyvinyl)tin
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Indicaciones de peligro
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.