Learn More
Transparent Glass Staining Trough
Product Code. 12888735
Change view
Click to view available options
Type:
Staining Jar
Height (Metric):
50 mm
Unit Size:
Each
Product Code. Type Height (Metric) Unit Size
12888735 Staining Jar 50 mm Each
1 options
This item is not returnable. View return policy
Product Code. 12888735 Supplier Brand Not Defined Supplier No. 2960/1
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Prevent evaporation of staining liquid using the drop-on lid supplied with the Transparent Glass Staining Trough. This heavy glass staining jar is an essential microscope accessory for the laboratory.
  • Glass trough with lid
  • Holds microscope slides 76mm x 26mm
  • Two different styles
Specifications
Closure Included Closure Included
Bottom Shape Flat
Material Soda Lime Glass
Type Staining Jar
For Use With (Application) Staining Rack
Slide Count 25
Dimensions (L x W) 112 x 112 mm
Dimensions (L x W x H) 112 x 112 x 50 mm
Length (Metric) 112 mm
Width (Metric) 112 mm
Height (Metric) 50 mm
Depth (Metric) 112 mm